accessibility menu, dialog, popup

UserName

CAS RN 36327-91-0

H2OOONHOHN

CAS RN 36327-91-0

IUPAC Name: 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carbaldehyde hydrate
Synonyms: 5-formyluracil hydrate
Molecular Weight (g/mol): 158.11
Molecular Formula: C5H6N2O4
InChi Key: FFSRLRNGSZKHTN-UHFFFAOYSA-N
SMILES: O.O=CC1=CC(=O)NC(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results