Learn More
Fisherbrand™ Upright ULT Freezer Racks for Microtiter Plates
Product Code. 15729559 Shop All Fisherbrand Products
Change view
Click to view available options
Holds:
Plates with Lid (70)
Plates with Lid (84)
Plates with Lid (96)
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
15729559 Plates with Lid (70) Each
15739559 Plates with Lid (84) Each
15749559 Plates with Lid (96) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15729559
Only null left
Add to basket
Add to basket
Fisherbrand inventory control racks for 96-Well and 384-Well microtiter plates. They fit all standard types of upright ULT freezers.
  • Holds up to 96 plates with lid or 120 plates without lid
  • Made of high quality stainless steel
  • Side opening with handles on the front for easy plate retrieval.
Specifications
Holds Plates with Lid (70)
Material Stainless Steel
For Use With (Application) 96/384-Well Microtiter Plates
For Use With (Equipment) Upright ULT Freezer
Dimensions (L x W x H) 466 x 233 x140 mm
Product Type Side Access Freezer Rack