missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UPF3B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57233
This item is not returnable.
View return policy
Description
UPF3B Polyclonal specifically detects UPF3B in Human samples. It is validated for Western Blot.
Specifications
| UPF3B | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| HUPF3B, Nonsense mRNA reducing factor 3B, regulator of nonsense transcripts 3B, RENT3BhUpf3p-X, UPF3 regulator of nonsense transcripts homolog B (yeast), UPF3XMRXS14, Up-frameshift suppressor 3 homolog B, Up-frameshift suppressor 3 homolog on chromosome X | |
| Rabbit | |
| 58 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Mouse: 92%; Rat: 92%. | |
| Human, Mouse, Rat, Pig, Canine, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9BZI7 | |
| UPF3B | |
| Synthetic peptides corresponding to UPF3B(UPF3 regulator of nonsense transcripts homolog B (yeast)) The peptide sequence was selected from the middle region of UPF3B (NP_542199) Peptide sequence KIDRIPERDKLKDEPKIKVHRFLLQAVNQKNLLKKPEKGDEKELDKREKA. | |
| Affinity purified | |
| RUO | |
| 65109 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction