Learn More
UPF1 Mouse, anti-Human, CoraLite Plus 488, Clone: 3B6B5, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | UPF1 |
|---|---|
| Clone | 3B6B5 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18004715
|
Proteintech
CL488-66898-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18058955
|
Proteintech
CL488-66898-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The UPF1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UPF1. This antibody recognizes human, mouse, rat antigen. The UPF1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
| UPF1 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 5976 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 3B6B5 | |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human, Mouse, Rat | |
| ATP dependent helicase RENT1, FLJ43809, FLJ46894, HUPF1, KIAA0221, NORF1, pNORF1, RENT1, UPF1 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.