Learn More
UPF1 Mouse, anti-Human, CoraLite Plus 488, Clone: 3B6B5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66898-100UL
Description
The UPF1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UPF1. This antibody recognizes human, mouse, rat antigen. The UPF1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
| UPF1 | |
| 3B6B5 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| ATP dependent helicase RENT1, FLJ43809, FLJ46894, HUPF1, KIAA0221, NORF1, pNORF1, RENT1, UPF1 | |
| 100 μL | |
| 5976 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |