missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uPAR Antibody (IIIB6), Novus Biologicals™
Mouse Monoclonal Antibody
£186.00 - £558.00
Specifications
| Antigen | uPAR |
|---|---|
| Clone | IIIB6 |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18289443
|
Novus Biologicals
NBP2-62789 |
100 μg |
£558.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18253035
|
Novus Biologicals
NBP2-62789-0.025MG |
0.025 mg |
£186.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
uPAR Monoclonal specifically detects uPAR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| uPAR | |
| Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| Cancer | |
| PBS with 0.1% BSA with 0.02% Sodium Azide | |
| 5329 | |
| uPAR (D1 domain) | |
| Primary |
| IIIB6 | |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
| PLAUR | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title