missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uPAR Antibody (IIIB6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62789-0.025MG
This item is not returnable.
View return policy
Description
uPAR Monoclonal specifically detects uPAR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| uPAR | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.1% BSA with 0.02% Sodium Azide | |
| PLAUR | |
| uPAR (D1 domain) | |
| 0.025 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| IIIB6 | |
| Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
| Mouse | |
| Protein G purified | |
| Cancer | |
| 5329 | |
| Store at 4degreesC. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction