Learn More
Corning™ Universal Fit Pipet Tips: Racked, Sterile
Designed to assure a reliable fit with major brand pipetters
Brand:  Corning™ 9032
Code : 66
Additional Details : Weight : 2.07000kg
 View more versions of this product
Product Code. 10266721
  • £98.36 / Pack of 1000
Estimated Shipment: 20-09-2024
to see stock.
Add to basket
Description
Description
  • Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
  • Beveled orifice helps ensure accurate fluid delivery.
  • 1-200 μL universal fit tips are graduated at the 250 μL, 500 μL, and 1,000 μL volumes.
  • Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
  • RNase-/DNase-free.
  • Nonpyrogenic.
Specifications
Specifications
100 to 1000 μL
Blue
Round
Yes
Sterile
Rack
Rack
Polypropylene