missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Universal Fit Pipet Tips: Racked, Sterile
GREENER_CHOICE

Product Code. p-8015610 Shop All Corning Products
Click to view available options
Volume (Metric):
1 to 200 μL
100 to 1000 μL
Color:
Blue
Natural
Yellow
Unit Size:
Pack of 1000
Pack of 960
This item is not returnable. View return policy

Product Code. 10649321

Brand: Corning™ 4864

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed to assure a reliable fit with major brand pipetters

  • Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
  • Beveled orifice helps ensure accurate fluid delivery.
  • 1-200 μL universal fit tips are graduated at the 250 μL, 500 μL, and 1,000 μL volumes.
  • Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
  • RNase-/DNase-free.
  • Nonpyrogenic.
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Sterility Sterile
Color Natural
Format Rack
Tip Style Round
Packaging Rack
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.