missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ UNISOL pH Indicator Solution

Product Code. 11356554 Shop All Macherey-Nagel Products
Click to view available options
pH Range:
4 to 10
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 11356554

Brand: Macherey-Nagel™ 91002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For pH determination in pure water, in surface waters, and in very dilute acids or bases.

  • pH determinations in weakly buffered solutions
    • UNISOL indicator solutions are used for pH determination in pure water, in surface waters and in very dilute acids or bases.
    • An indicator solution is added to the sample and the reaction color is compared with a color scale.
    • Therefore, they allow the easy and reliable pH determination in weakly buffered solutions.

Specifications

pH Range 4 to 10
Volume (Metric) 100 mL
Includes Color chart, cuvette
Quantity 1 Bottle
Type pH Indicator Solution
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.