missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC5CL Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13322-1-AP-20UL
This item is not returnable.
View return policy
Description
UNC5CL is a membrane protein belonging to theUNC-5 family with a death domain and a ZU5 domain and is known to interact with p65/RELA and NFKB1. UNC5CL is known to interact with subunit p105 of NF-kappaB and transactivator p65 and is thus involved in inhibition of NF-kappaB activation and NF-kappaB-dependent transcription by inhibiting the binding of NF-kappaB to its target sequence (providing an alternative regulatory mechanism for NF-kappaB-mediated transcription). This regulatory protein sensitizes cells to apoptosis induced by TNF and TRAIL.Specifications
| UNC5CL | |
| Polyclonal | |
| Unconjugated | |
| UNC5CL | |
| Protein unc 5 homolog C like, UNC5C like protein, UNC5CL, ZUD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 222643, 76589 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6R653, Q8IV45 | |
| Unc5cl | |
| UNC5CL Fusion Protein Ag3970 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction