Learn More
UNC13D/Munc13-4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16905-1-AP-20UL
 View more versions of this product
Product Code. 16816674
  • £135.00 / 20µL
Description
This gene encodes a protein that is a member of the UNC13 family, containing similar domain structure as other family members but lacking an N-terminal phorbol ester-binding C1 domain present in other Munc13 proteins. The protein appears to play a role in vesicle maturation during exocytosis and is involved in regulation of cytolytic granules secretion. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis type 3, a genetically heterogeneous, rare autosomal recessive disorder.
Specifications
Specifications
UNC13D/Munc13-4
Polyclonal
Unconjugated
UNC13D
FHL3, HLH3, HPLH3, Munc13 4, Munc13-4, Protein unc 13 homolog D, unc 13 homolog D (C. elegans), UNC13D
Rabbit
Antigen Affinity Chromatography
RUO
201294
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q70J99
UNC13D
UNC13D/Munc13-4 Fusion Protein Ag10519
20 μL