missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC13D/Munc13-4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | UNC13D/Munc13-4 |
|---|---|
| Concentration | 0.23 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16816674
|
Proteintech
16905-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16806674
|
Proteintech
16905-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a protein that is a member of the UNC13 family, containing similar domain structure as other family members but lacking an N-terminal phorbol ester-binding C1 domain present in other Munc13 proteins. The protein appears to play a role in vesicle maturation during exocytosis and is involved in regulation of cytolytic granules secretion. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis type 3, a genetically heterogeneous, rare autosomal recessive disorder.Specifications
| UNC13D/Munc13-4 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q70J99 | |
| 201294 | |
| UNC13D/Munc13-4 Fusion Protein Ag10519 | |
| Primary | |
| -20°C | |
| UNC13D |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| FHL3, HLH3, HPLH3, Munc13 4, Munc13-4, Protein unc 13 homolog D, unc 13 homolog D (C. elegans), UNC13D | |
| UNC13D | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title