missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UMP/CMP kinase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | UMP/CMP kinase |
|---|---|
| Concentration | 0.23 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16828383
|
Proteintech
11360-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16818383
|
Proteintech
11360-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UCK1 (Uridine-cytidine kinase 1) phosphorylates uridine and cytidine to uridine monophosphate and cytidine monophosphate. This enzyme does not phosphorylate deoxyribonucleosides or purine ribonucleosides. Of note, UCK1 is able to use either ATP or GTP as a phosphate donor. UCK1 also possesses the ability to phosphorylate a number of cytidine and uridine nucleoside analogs such as 6-azauridine, 5-fluorouridine, 4-thiouridine, 5-bromouridine, N(4)-acetylcytidine, N(4)-benzoylcytidine, 5-fluorocytidine, 2-thiocytidine, 5-methylcytidine, and N(4)-anisoylcytidine.Specifications
| UMP/CMP kinase | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| P30085, Q4KM73, Q9DBP5 | |
| 298410, 51727, 66588 | |
| UMP/CMP kinase Fusion Protein Ag1920 | |
| Primary | |
| -20°C | |
| CMPK1 |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CMK, CMPK, CMPK1, Cytidine monophosphate kinase, Cytidylate kinase, Deoxycytidylate kinase, UCK, UMK, UMP CMP kinase, UMP CMPK, UMP/CMP kinase, UMP/CMPK, UMPK, Uridine monophosphate kinase | |
| CMPK1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title