missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UMP/CMP kinase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11360-1-AP-20UL
This item is not returnable.
View return policy
Description
UCK1 (Uridine-cytidine kinase 1) phosphorylates uridine and cytidine to uridine monophosphate and cytidine monophosphate. This enzyme does not phosphorylate deoxyribonucleosides or purine ribonucleosides. Of note, UCK1 is able to use either ATP or GTP as a phosphate donor. UCK1 also possesses the ability to phosphorylate a number of cytidine and uridine nucleoside analogs such as 6-azauridine, 5-fluorouridine, 4-thiouridine, 5-bromouridine, N(4)-acetylcytidine, N(4)-benzoylcytidine, 5-fluorocytidine, 2-thiocytidine, 5-methylcytidine, and N(4)-anisoylcytidine.Specifications
| UMP/CMP kinase | |
| Polyclonal | |
| Unconjugated | |
| CMPK1 | |
| CMK, CMPK, CMPK1, Cytidine monophosphate kinase, Cytidylate kinase, Deoxycytidylate kinase, UCK, UMK, UMP CMP kinase, UMP CMPK, UMP/CMP kinase, UMP/CMPK, UMPK, Uridine monophosphate kinase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 298410, 51727, 66588 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P30085, Q4KM73, Q9DBP5 | |
| CMPK1 | |
| UMP/CMP kinase Fusion Protein Ag1920 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction