missing translation for 'onlineSavingsMsg'
Learn More

Elma™ Ultrasonic™ Glass Beaker

Product Code. 10228811
Click to view available options
Capacity (Metric):
1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Perfect for containing small parts for ultrasonic cleaning. Elma™ Ultrasonic™ Glass Beaker can be used to conserve ultrasonic cleaning solution.

  • Useful for ultrasonic cleaning of small parts.
  • Conserves ultrasonic cleaning solution.
  • Beakers containing cleaning parts and cleaning solution are suspended in a water bath containing surfactant.

Specifications

Color Clear
Diameter (Metric) Outer 95 mm
Capacity (Metric) 1000 mL
Material Glass
For Use With (Application) Elmasonic Ultrasonic Bath
Type Beaker
Graduated Yes
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'