Learn More
Thermo Scientific™ UltraLink™ Iodoacetyl Micro Peptide Coupling Kit, 0.1 mL
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ UltraLink Iodoacetyl Resin and Peptide Coupling Kit enable the covalent immobilization of cysteine-containing peptides for use in antibody purification and other affinity protocols.
The Micro Peptide Coupling Kit is a microcentrifuge spin column kit for immobilizing small amounts (25 to 250μg) of sulfhydryl-containing peptides (e.g., cysteine-terminated peptides) onto the resin to create a small, reusable affinity column. The coupling and affinity purification procedures are optimized for small sample volumes (200 to 300μL). Wash and elution steps are achieved rapidly and efficiently with the convenient microcentrifuge spin columns. Each kit contains sufficient reagents for 10 coupling reactions and 20 affinity purifications. The kit is ideal for immobilizing peptide antigens that contain a terminal cysteine residue for use in purifying specific antibodies from small serum, ascites or culture supernatant samples.
Highlights:
High coupling efficiency and capacity – immobilizes sulfhydryl- containing proteins or other ligands with high efficiency and coupling capacity in one hour
Specific and leak-proof coupling chemistry – reacts specifically with sulfhydryl groups (reduced thiols), resulting in thioether bonds that are stable for use in many affinity purification procedures
Simple coupling conditions – perform immobilization reaction in any of a variety of buffers; coupling is most efficient and specific at pH 8.0 to 8.5; coupling compatible with or without organic solvent
Excellent reusability – prepared affinity resin can be used with typical binding and elution procedures for many cycles of affnity purification without significant loss of binding capacity
Durable, high-performance resin – porous beads have a 60μm diameter, can withstand 100 psi (6.9 bar) and allow for linear flow-through rates of 3000cm/hour
Micro Peptide Coupling Kit (Part No. 20485):
Optimized for small scale – ideal for coupling small amounts (25 to 250μg) of sulfhydryl-containing peptide or protein for purification of specific antibodies from crude serum
High-performance affinity resin – uses durable, polyacrylamide-based UltraLink Iodoacetyl Resin for specific reaction to sulfhydryl groups
Efficient coupling chemistry – immobilization efficiency >85% (as measured with 1 hour reaction using insulin, calcitonin and osteocalcin peptides)
Fast and easy to use – perform wash and elution steps using a microcentrifuge
Reusable – use prepared peptide resin several times with no significant loss of capacity
Includes:
Micro Peptide Coupling Kit contains microcentrifuge spin columns containing UltraLink Iodoacetyl Resin, coupling buffer, L-cysteine (for quenching), wash solution, PBS, elution buffer and collection tubes
Specifications
Specifications
| Capacity (Metric) | 25 to 250 μg of Peptide or Protein per Reaction |
| Description | UltraLink Iodoacetyl Micro Peptide Coupling Kit |
| Content And Storage | Sufficient For: Immobilizing 25–250 μg of peptide or protein per microcentrifuge column; Includes buffers for 20 affinity purifications • UltraLink Iodoacetyl Spin Columns, 0.1 mL, 10 columns • Coupling Buffer, 100 mL • L-Cysteine·HCl, 100 mg • Wash Solution, 25 mL • PBS Pack (makes 500 mL), 1 pack • IgG Elution Buffer, 50 mL • Collection Tubes, 200 tubes Store at 4°C protected from light. |
| Product Line | UltraLink |
| Quantity | 1 Kit, 10 Columns |
| Type | Peptide Coupling Kit |
| Stationary Phase | Haloacetyl |
| Sufficient For | 10 Reactions |
| Format | Kit, Microcentrifuge, Spin Column |
| Column Type | Affinity, UltraLink™ Resin |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.