Learn More
Buerkle™ Ultraflex Safety Goggles
Product Code. 11971651 Shop All Bürkle Products
Change view
Click to view available options
Frame Color:
Blue
Unit Size:
Each
Product Code. Frame Color Unit Size
11971651 Blue Each
1 options
Product Code. 11971651 Supplier Buerkle™ Supplier No. 25021003
  • ONLINE EXCLUSIVE PRICE
    £39.70
    £37.61 / Pack of 1
    Save £2.09
    5% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Hingeless side pieces and soft side piece ends
  • Ideal as over-glasses for conventional prescription glasses
  • Ultralight for maximum wearing comfort
  • Safety spectacles without hinges, completely metal-free
  • Optimum optical clarity
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DIN EN 166 - 168 and 170
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Over-the-Glasses Safety Glasses
For Use With (Application) Eye Protection
Frame Color Blue
No. per Pack 1 Ea.
Material Polycarbonate
Disposable Limited-use
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400