missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK3 Rabbit anti-Human, Clone: S08-7J3, Novus Biologicals™
Rabbit Monoclonal Antibody
£224.00 - £451.00
Specifications
| Antigen | ULK3 |
|---|---|
| Clone | S08-7J3 |
| Dilution | Western Blot, ELISA, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, Immunofluorescence |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18343695
|
Novus Biologicals
NBP3-15108-25UL |
25 μg |
£224.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18357044
|
Novus Biologicals
NBP3-15108-100UL |
100 μg |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ULK3 Monoclonal antibody specifically detects ULK3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ImmunofluorescenceSpecifications
| ULK3 | |
| Western Blot, ELISA, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZP434C131, EC 2.7.11.1, FLJ90566, serine/threonine-protein kinase ULK3, Unc-51-like kinase 3, unc-51-like kinase 3 (C. elegans) | |
| A synthetic peptide of human ULK3 (Uniprot#: Q6PHR2) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S08-7J3 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 25989 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title