missing translation for 'onlineSavingsMsg'
Learn More

ULK3 Rabbit anti-Human, Clone: S08-7J3, Novus Biologicals™

Product Code. 18343695 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343695 25 μg 25µL
18357044 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18343695 Supplier Novus Biologicals Supplier No. NBP31510825UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ULK3 Monoclonal antibody specifically detects ULK3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ULK3
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone S08-7J3
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, Immunofluorescence
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias DKFZP434C131, EC 2.7.11.1, FLJ90566, serine/threonine-protein kinase ULK3, Unc-51-like kinase 3, unc-51-like kinase 3 (C. elegans)
Host Species Rabbit
Immunogen A synthetic peptide of human ULK3 (Uniprot#: Q6PHR2)
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 25989
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.