missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ULK2, GST Tag Recombinant Protein
Human ULK2, GST Tag Recombinant Protein
£431.00 - £6530.00
Specifications
Gene Alias | Unc51.2, KIAA0623 |
---|---|
Protein Length | aa 1-478. Contains V370M SNP |
Species | Human |
Recombinant | Recombinant |
Protein Tag | GST Tag |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15429053
|
Invitrogen™
PV6434 |
100μg |
£1460.00
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15441234
|
Invitrogen™
PV6433 |
10μg |
£431.00
10µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15439053
|
Invitrogen™
PV6435 |
1mg |
£6530.00
1mg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Mammalian Atg13 binds ULK2 and mediates the interaction of the ULK protein with FIP200. The binding of Atg13 stabilizes and activates ULK and facilitates the phosphorylation of FIP200 by ULK. The ULK-Atg13-FIP200 complex is a direct target of mTOR and important regulator of autophagy in response to mTOR signalling. Yeast 2-hybrid assays showed the C-terminus of ULK2 binds the C-terminus of SynGAP, a negative regulator of Ras that is associated with neural development.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Unc51.2, KIAA0623 | |
Human | |
GST Tag |
aa 1-478. Contains V370M SNP | |
Recombinant |