Learn More
Invitrogen™ Human ULK2, GST Tag Recombinant Protein
Human ULK2, GST Tag Recombinant Protein
Brand: Invitrogen™ PV6433
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Mammalian Atg13 binds ULK2 and mediates the interaction of the ULK protein with FIP200. The binding of Atg13 stabilizes and activates ULK and facilitates the phosphorylation of FIP200 by ULK. The ULK-Atg13-FIP200 complex is a direct target of mTOR and important regulator of autophagy in response to mTOR signalling. Yeast 2-hybrid assays showed the C-terminus of ULK2 binds the C-terminus of SynGAP, a negative regulator of Ras that is associated with neural development.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Kinase Assay | |
10μg | |
aa 1-478. Contains V370M SNP | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.25 mM DTT, 10 mM glutathione, 0.1 mM EDTA, 0.1 mM PMSF, 25% glycerol Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. |
|
Serine/Threonine Kinase | |
Baculovirus | |
Recombinant, Fragment | |
Serine/Threonine Kinases |
81.3 kDa | |
Unc51.2, KIAA0623 | |
Human | |
GST Tag | |
Biochemical binding/kinetic | |
ULK2 | |
Kinases & Inhibitors | |
Liquid |