missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00 - £348.00
Specifications
| Antigen | ULK1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18363006
|
Novus Biologicals
NBP3-16957-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18304133
|
Novus Biologicals
NBP3-16957-100UL |
100 μg |
£348.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ULK1 Polyclonal antibody specifically detects ULK1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| ULK1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ATG1, ATG1 autophagy related 1 homolog, ATG1A, EC 2.7.11, EC 2.7.11.1, FLJ38455, FLJ46475, KIAA0722, serine/threonine-protein kinase ULK1, UNC51, unc-51 (C. elegans)-like kinase 1, Unc51.1, Unc-51-like kinase 1, unc-51-like kinase 1 (C. elegans) | |
| A synthetic phosphorylated peptide around S467 of human ULK1 (NP_003556.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Autophagy, mTOR Pathway, Protein Kinase | |
| PBS, 50% glycerol, pH7.3 | |
| 8408 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title