missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-16957-100UL
This item is not returnable.
View return policy
Description
ULK1 Polyclonal antibody specifically detects ULK1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ULK1 | |
| Polyclonal | |
| PBS, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| ATG1, ATG1 autophagy related 1 homolog, ATG1A, EC 2.7.11, EC 2.7.11.1, FLJ38455, FLJ46475, KIAA0722, serine/threonine-protein kinase ULK1, UNC51, unc-51 (C. elegans)-like kinase 1, Unc51.1, Unc-51-like kinase 1, unc-51-like kinase 1 (C. elegans) | |
| A synthetic phosphorylated peptide around S467 of human ULK1 (NP_003556.2). | |
| 100 μg | |
| Autophagy, mTOR Pathway, Protein Kinase | |
| 8408 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction