missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCP3 Antibody (6B8C6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | UCP3 |
|---|---|
| Clone | 6B8C6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18279562
|
Novus Biologicals
NBP2-61821 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692679
|
Novus Biologicals
NBP2-61821-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UCP3 Monoclonal specifically detects UCP3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| UCP3 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| mitochondrial uncoupling protein 3, SLC25A9Solute carrier family 25 member 9, UCP 3, uncoupling protein 3 (mitochondrial, proton carrier), Uncoupling protein-3 | |
| UCP3 | |
| IgG2a | |
| Protein G purified |
| 6B8C6 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| 7352 | |
| Purified recombinant fragment of human UCP3 (AA: 1-113 and 217-312) expressed in E. Coli. | |
| Primary | |
| 34.2 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title