missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCP3 Antibody (6B8C6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61821
This item is not returnable.
View return policy
Description
UCP3 Monoclonal specifically detects UCP3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| UCP3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| mitochondrial uncoupling protein 3, SLC25A9Solute carrier family 25 member 9, UCP 3, uncoupling protein 3 (mitochondrial, proton carrier), Uncoupling protein-3 | |
| Mouse | |
| 34.2 kDa | |
| 0.1 mL | |
| 7352 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 6B8C6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| UCP3 | |
| Purified recombinant fragment of human UCP3 (AA: 1-113 and 217-312) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction