missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCH-L1/PGP9.5 Antibody (UCHL1/775), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44795-0.02MG
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry (Paraffin)
UCH-L1/PGP9.5 Monoclonal specifically detects UCH-L1/PGP9.5 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| UCH-L1/PGP9.5 | |
| Monoclonal | |
| 0.2mg/mL | |
| Western Blot 0.5 - 1.0 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, SDS-Page | |
| P09936 | |
| UCHL1 | |
| Recombinant human UCHL1 protein | |
| 0.02 mg | |
| Alzheimers Research, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 7345 | |
| Human, Rat | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), SDS-Page | |
| UCHL1/775 | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This MAb reacts with a protein of 20-30kDa, identified as PGP9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UchL1). Initially, PGP9.5 expression in normal tissues was reported in neurons and neuroendocrine cells but later it was found in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. Furthermore, immunostaining for PGP9.5 has been shown in a wide variety of mesenchymal neoplasms as well. A mutation in PGP9.5 gene is believed to cause a form of Parkinson's disease. | |
| Store at 4C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction