missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBXN10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26062-1-AP-20UL
This item is not returnable.
View return policy
Description
UBXN10 (VCP/p97) binding protein required for ciliogenesis. Acts as a tethering factor that facilitates recruitment of VCP/p97 to the intraflagellar transport complex B (IFT-B) in cilia. UBX domain-containing proteins act as tethering factors for VCP/p97 and may specify substrate specificity of VCP/p97.Specifications
| UBXN10 | |
| Polyclonal | |
| Unconjugated | |
| UBXN10 | |
| UBX domain protein 10, UBXD3, UBXN10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 127733, 212190 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BG34, Q96LJ8 | |
| Ubxn10 | |
| UBXN10 Fusion Protein Ag22037 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction