missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBXN10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | UBXN10 |
|---|---|
| Concentration | 0.28 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16804595
|
Proteintech
26062-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16894585
|
Proteintech
26062-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UBXN10 (VCP/p97) binding protein required for ciliogenesis. Acts as a tethering factor that facilitates recruitment of VCP/p97 to the intraflagellar transport complex B (IFT-B) in cilia. UBX domain-containing proteins act as tethering factors for VCP/p97 and may specify substrate specificity of VCP/p97.Specifications
| UBXN10 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q8BG34, Q96LJ8 | |
| 127733, 212190 | |
| UBXN10 Fusion Protein Ag22037 | |
| Primary | |
| -20°C | |
| UBXN10 |
| 0.28 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| UBX domain protein 10, UBXD3, UBXN10 | |
| Ubxn10 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.