missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ubiquitin E1 Inhibitor, PYR-41

Product Code. p-7082459
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 15416879

Brand: Thermo Scientific Chemicals J65388.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 418805-02-4 | C17H13N3O7·3H2O, C17H13N3O7 | 371.31 g/mol

PYR 41 is an irreversible and cell-permeable UBE1 inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • PYR-41 is a cell permeable inhibitor of Ubiquitin activating enzyme (E1) with little or no activity against E3, E2, or caspase enzymatic activity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 418805-02-4
Molecular Formula C17H13N3O7·3H2O, C17H13N3O7
Synonym Ubiquitin-Activating Enzyme E1 Inhibitor; PYR-41

Specifications

Quantity 100 mg
Description Dark Purple
Content And Storage -30°C to -10°C
For Use With (Application) PYR-41 is a cell permeable inhibitor of Ubiquitin activating enzyme (E1) with little or no activity against E3, E2, or caspase enzymatic activity.
Formula Weight 425.40g/mol
Sensitivity Light Sensitive
Solubility Soluble in DMSO at 5mg/ml with warming
Product Type Signal Transduction Reagent
Form Solid
InChI Key ARGIPZKQJGFSGQ-UHFFFAOYSA-N
SMILES CCOC(=O)C1=CC=C(C=C1)N1NC(=O)C(=CC2=CC=C(O2)[N+]([O-])=O)C1=O
IUPAC Name ethyl 4-{4-[(5-nitrofuran-2-yl)methylidene]-3,5-dioxopyrazolidin-1-yl}benzoate
Molecular Weight (g/mol) 371.31
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.