missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Ubiquitin E1 Inhibitor, PYR-41
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
PYR 41 is an irreversible and cell-permeable UBE1 inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- PYR-41 is a cell permeable inhibitor of Ubiquitin activating enzyme (E1) with little or no activity against E3, E2, or caspase enzymatic activity
Specifications
Specifications
| Quantity | 25 mg |
| Description | Dark Purple |
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | PYR-41 is a cell permeable inhibitor of Ubiquitin activating enzyme (E1) with little or no activity against E3, E2, or caspase enzymatic activity. |
| Formula Weight | 425.40g/mol |
| Sensitivity | Light Sensitive |
| Solubility | Soluble in DMSO at 5mg/ml with warming |
| Product Type | Signal Transduction Reagent |
| Form | Solid |
| InChI Key | ARGIPZKQJGFSGQ-UHFFFAOYSA-N |
| Show More |
Safety and Handling
Product Identifier
- Ubiquitin E1 Inhibitor, PYR-41
- Warning
- Acute toxicity Category 4
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : Ubiquitin E1 Inhibitor, PYR-41
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction