missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin-activating Enzyme/UBE1 Antibody (05), Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £255.00
Specifications
| Antigen | Ubiquitin-activating Enzyme/UBE1 |
|---|---|
| Clone | 05 |
| Dilution | ELISA 1:5000-1:10000, Immunoprecipitation 0.2-1 μg/mg of lysate |
| Applications | ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18604938
|
Novus Biologicals
NBP3-05789-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692897
|
Novus Biologicals
NBP3-05789-50ul |
50 μg |
£173.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ubiquitin-activating Enzyme/UBE1 Monoclonal antibody specifically detects Ubiquitin-activating Enzyme/UBE1 in Human samples. It is validated for ELISA, ImmunoprecipitationSpecifications
| Ubiquitin-activating Enzyme/UBE1 | |
| ELISA 1:5000-1:10000, Immunoprecipitation 0.2-1 μg/mg of lysate | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| A1S9T, A1S9T and BN75 temperature sensitivity complementing, A1ST, AMCX1, GXP1, MGC4781, POC20, POC20 centriolar protein homolog, Protein A1S9, SMAX2, UBA1, ubiquitin-activating enzyme E1 homolog A, UBA1A, UBE1A1S9, UBE1X, Ubiquitin-activating enzyme E1, ubiquitin-activating enzyme E1 (A1S9T and BN75 temperature sensitivitycomplementing), ubiquitin-like modifier activating enzyme 1, ubiquitin-like modifier-activating enzyme 1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Ubiquitin-activating Enzyme/UBE1 (Accession#: NP_003325.2; Ser2-Arg1058). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 05 | |
| ELISA, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cell Cycle and Replication, Chromatin Research, Ubiquitin Proteasome Pathway | |
| 0.2 um filtered solution in PBS | |
| 7317 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title