missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin-activating Enzyme/UBE1 Antibody (05), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05789-50ul
This item is not returnable.
View return policy
Description
Ubiquitin-activating Enzyme/UBE1 Monoclonal antibody specifically detects Ubiquitin-activating Enzyme/UBE1 in Human samples. It is validated for ELISA, Immunoprecipitation
Specifications
| Ubiquitin-activating Enzyme/UBE1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunoprecipitation | |
| 05 | |
| ELISA 1:5000-1:10000, Immunoprecipitation 0.2-1 μg/mg of lysate | |
| A1S9T, A1S9T and BN75 temperature sensitivity complementing, A1ST, AMCX1, GXP1, MGC4781, POC20, POC20 centriolar protein homolog, Protein A1S9, SMAX2, UBA1, ubiquitin-activating enzyme E1 homolog A, UBA1A, UBE1A1S9, UBE1X, Ubiquitin-activating enzyme E1, ubiquitin-activating enzyme E1 (A1S9T and BN75 temperature sensitivitycomplementing), ubiquitin-like modifier activating enzyme 1, ubiquitin-like modifier-activating enzyme 1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Ubiquitin-activating Enzyme/UBE1 (Accession#: NP_003325.2; Ser2-Arg1058). | |
| 50 μg | |
| Cell Biology, Cell Cycle and Replication, Chromatin Research, Ubiquitin Proteasome Pathway | |
| 7317 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction