missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2K/E2-25K Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£378.00
Specifications
| Applications | Western Blot |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Host Species | Rabbit |
Description
UBE2K/E2-25K Polyclonal specifically detects UBE2K/E2-25K in Human samples. It is validated for Western Blot.Specifications
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| P61086 | |
| UBE2K | |
| Primary | |
| 22 kDa |
| Polyclonal | |
| Purified | |
| RUO | |
| 3093 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title