missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2F/NCE2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | UBE2F/NCE2 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
UBE2F/NCE2 Polyclonal specifically detects UBE2F/NCE2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
| UBE2F/NCE2 | |
| Polyclonal | |
| Rabbit | |
| Q969M7 | |
| 140739 | |
| Synthetic peptides corresponding to UBE2F(ubiquitin-conjugating enzyme E2F (putative)) The peptide sequence was selected from the N terminal of UBE2F. Peptide sequence MLTLASKLKRDDGLKGSRTAATASDSTRRVSVRDKLLVKEVAELEANLPC. | |
| Primary |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| EC 6.3.2, EC 6.3.2.-, MGC18120, NCE2Ubiquitin-conjugating enzyme E2 F, NEDD8 carrier protein UBE2F, NEDD8 conjugating enzyme, NEDD8 protein ligase UBE2F, NEDD8-conjugating enzyme 2, NEDD8-conjugating enzyme UBE2F, ubiquitin-conjugating enzyme E2F (putative) | |
| UBE2F | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title