missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2F/NCE2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57602
This item is not returnable.
View return policy
Description
UBE2F/NCE2 Polyclonal specifically detects UBE2F/NCE2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
| UBE2F/NCE2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 6.3.2, EC 6.3.2.-, MGC18120, NCE2Ubiquitin-conjugating enzyme E2 F, NEDD8 carrier protein UBE2F, NEDD8 conjugating enzyme, NEDD8 protein ligase UBE2F, NEDD8-conjugating enzyme 2, NEDD8-conjugating enzyme UBE2F, ubiquitin-conjugating enzyme E2F (putative) | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 140739 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q969M7 | |
| UBE2F | |
| Synthetic peptides corresponding to UBE2F(ubiquitin-conjugating enzyme E2F (putative)) The peptide sequence was selected from the N terminal of UBE2F. Peptide sequence MLTLASKLKRDDGLKGSRTAATASDSTRRVSVRDKLLVKEVAELEANLPC. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Human: 100%; Rat: 100%; Xenopus: 92%; Canine: 92%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction