missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE1 Mouse, anti-Human, CoraLite Plus 488, Clone: 1H7E8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | UBE1 |
|---|---|
| Clone | 1H7E8 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18090346
|
Proteintech
CL488-67198-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18065195
|
Proteintech
CL488-67198-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The UBE1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UBE1. This antibody recognizes human, mouse, rat antigen. The UBE1 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| UBE1 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 7317 | |
| IgG2a | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1H7E8 | |
| Western Blot | |
| Liquid | |
| Human, Mouse, Rat | |
| A1S9, A1S9T, A1ST, AMCX1, GXP1, Protein A1S9, SMAX2, UBA1, UBA1A, UBE1, UBE1X, Ubiquitin activating enzyme E1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title