Learn More
UBE1 Mouse, anti-Human, CoraLite Plus 488, Clone: 1H7E8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67198-20UL
Description
The UBE1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UBE1. This antibody recognizes human, mouse, rat antigen. The UBE1 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| UBE1 | |
| 1H7E8 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 155 kDa | |
| IgG2a |
| Western Blot | |
| 1000 μg/mL | |
| A1S9, A1S9T, A1ST, AMCX1, GXP1, Protein A1S9, SMAX2, UBA1, UBA1A, UBE1, UBE1X, Ubiquitin activating enzyme E1 | |
| 20 μL | |
| 7317 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |