missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBA5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | UBA5 |
|---|---|
| Concentration | 0.25 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16859603
|
Proteintech
12093-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16849603
|
Proteintech
12093-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the E1-like ubiquitin-activating enzyme family. This protein activates ubiquitin-fold modifier 1, a ubiquitin-like post-translational modifier protein, via the formation of a high-energy thioester bond. Alternative splicing results in multiple transcript variants. A pseudogene of this gene has been identified on chromosome 1.Specifications
| UBA5 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q8VE47, Q9GZZ9 | |
| 66663, 79876 | |
| UBA5 Fusion Protein Ag2734 | |
| Primary | |
| -20°C | |
| UBA5 |
| 0.25 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| FLJ23251UBA5, THIFP1, UBA5, UBE1DC1, Ubiquitin activating enzyme 5, UFM1 activating enzyme | |
| Uba5 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title