missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBA5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12093-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the E1-like ubiquitin-activating enzyme family. This protein activates ubiquitin-fold modifier 1, a ubiquitin-like post-translational modifier protein, via the formation of a high-energy thioester bond. Alternative splicing results in multiple transcript variants. A pseudogene of this gene has been identified on chromosome 1.Specifications
| UBA5 | |
| Polyclonal | |
| Unconjugated | |
| UBA5 | |
| FLJ23251UBA5, THIFP1, UBA5, UBE1DC1, Ubiquitin activating enzyme 5, UFM1 activating enzyme | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 66663, 79876 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8VE47, Q9GZZ9 | |
| Uba5 | |
| UBA5 Fusion Protein Ag2734 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction