missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UACA Antibody (AE-5), Novus Biologicals™
Mouse Monoclonal Antibody
£196.00 - £377.00
Specifications
| Antigen | UACA |
|---|---|
| Clone | AE-5 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 μg/ml, Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence 1-2 μg/ml |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18628316
|
Novus Biologicals
NBP3-11313-100ug |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606708
|
Novus Biologicals
NBP3-11313-20ug |
20 μg |
£196.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UACA Monoclonal antibody specifically detects UACA in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| UACA | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis | |
| 10 mM PBS with 0.05% BSA | |
| 55075 | |
| IgG1 κ | |
| Protein A or G purified |
| AE-5 | |
| Western Blot 1-2 μg/ml, Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence 1-2 μg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| FLJ10128, KIAA1561MGC141969, MGC141967, nuclear membrane binding protein, NUCLING, uveal autoantigen with coiled coil domains and ankyrin repeats, uveal autoantigen with coiled-coil domains and ankyrin repeats | |
| Nuclei of myeloid leukemia biopsy cells | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title