missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UACA Antibody (AE-5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11313-20ug
This item is not returnable.
View return policy
Description
UACA Monoclonal antibody specifically detects UACA in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| UACA | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 μg/ml, Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence 1-2 μg/ml | |
| FLJ10128, KIAA1561MGC141969, MGC141967, nuclear membrane binding protein, NUCLING, uveal autoantigen with coiled coil domains and ankyrin repeats, uveal autoantigen with coiled-coil domains and ankyrin repeats | |
| Nuclei of myeloid leukemia biopsy cells | |
| 20 μg | |
| Apoptosis | |
| 55075 | |
| Store at 4°C. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| AE-5 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction