missing translation for 'onlineSavingsMsg'
Learn More
Learn More
U2AF2 Rabbit anti-Human, Mouse, Rat, Clone: 8E8R9, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £425.00
Specifications
| Antigen | U2AF2 |
|---|---|
| Clone | 8E8R9 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18379236
|
Novus Biologicals
NBP3-16432-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18357445
|
Novus Biologicals
NBP3-16432-100UL |
100 μg |
£425.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
U2AF2 Monoclonal antibody specifically detects U2AF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| U2AF2 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 11338 | |
| IgG | |
| Affinity purified |
| 8E8R9 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| hU2AF(65), hU2AF65, splicing factor U2AF 65 kD subunit, splicing factor U2AF 65 kDa subunit, U2 (RNU2) small nuclear RNA auxiliary factor 2, U2 auxiliary factor 65 kDa subunit, U2 small nuclear ribonucleoprotein auxiliary factor (65kD), U2 small nuclear RNA auxiliary factor 2, U2AF65U2 snRNP auxiliary factor large subunit | |
| A synthetic peptide corresponding to a sequence within amino acids 376-475 of human U2AF2/U2AF2 (P26368). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title