missing translation for 'onlineSavingsMsg'
Learn More

U2AF2 Rabbit anti-Human, Mouse, Rat, Clone: 8E8R9, Novus Biologicals™

Product Code. 18379236 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379236 20 μg 20µL
18357445 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18379236 Supplier Novus Biologicals Supplier No. NBP31643220UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

U2AF2 Monoclonal antibody specifically detects U2AF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen U2AF2
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8E8R9
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias hU2AF(65), hU2AF65, splicing factor U2AF 65 kD subunit, splicing factor U2AF 65 kDa subunit, U2 (RNU2) small nuclear RNA auxiliary factor 2, U2 auxiliary factor 65 kDa subunit, U2 small nuclear ribonucleoprotein auxiliary factor (65kD), U2 small nuclear RNA auxiliary factor 2, U2AF65U2 snRNP auxiliary factor large subunit
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 376-475 of human U2AF2/U2AF2 (P26368).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11338
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.