missing translation for 'onlineSavingsMsg'
Learn More

U0126, 99+%, Thermo Scientific Chemicals

Catalog No. 15413859
Ansicht ändern
Click to view available options
Quantity:
25 mg
100 mg
300 mg
Packungsgröße:
100mg
25mg
300mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packungsgröße
15413859 25 mg 25mg
15423859 100 mg 100mg
15403859 300 mg 300mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Catalog No. 15413859 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. J61246.MB
  • ONLINE EXCLUSIVE PRICE
    £162.18
    £134.65 / 25mg
    Save £27.53
    16% Off
Alle Werbeaktionen anzeigen
 Restricted Item
Estimated Shipment: 02-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Selective inhibitor of mitogen-activated protein kinases MEK-1 and MEK-2 | CAS: 109511-58-2 | C18H16N6S2, C18H16N6S2 | 380.49 g/mol

U0126, is used as a selective MAP Kinase Kinase (MKK, MEK) inhibitor. U-0126 has been observed to inhibit promoters containing an AP-1 response element, while in contrast having no effect on promoters that lack an AP-1 response group. U-0126 is also useful in neuronal studies in mice since it displays inhibitory effects against oxidative stress. U-0126 is an activator of PGC-1, mtTFA and nuclear respiratory factor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 109511-58-2
Molecular Formula C18H16N6S2
Synonym 1,4-Diamino-2,3-dicyano-1,4-bis(2-aminophenylthio)butadiene

Spezifikation

Quantity 25 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) Selective inhibitor of mitogen-activated protein kinases MEK-1 and MEK-2
Formula Weight 380.49g/mol
Solubility Soluble in DMSO; Poorly soluble in ethanol and water
Product Type Signal Transduction Reagent
Form Powder
Assay ≥99%
What is U0126, ≥99% (Cat. No. J61246.MC, J61246.MB) soluble in?

U0126, ≥99% (Cat. No. J61246.MC, J61246.MB) is soluble in DMSO. It is poorly soluble in ethanol and water.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.