missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals U-73122, 95%

Product Code. 15465189
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465189 1 mg 1mg
15455189 5 mg 5mg
15475189 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15465189 Supplier Thermo Scientific Chemicals Supplier No. J62898.M^
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 112648-68-7 | C29H40N2O3, C29H40N2O3 | 464.65 g/mol

It is used as a phospholipase C, phospholipase A2, and 5-LO inhibitor. It is also determined that in SK-N-SH neuroblastoma cells, U-73122 inhibits agonist-induced down-regulation of muscarinic receptors. In addition, it is a useful tool to investigate receptor-mediated PI turnover in signal transduction. U-73122 is a potent inhibitor of human neutrophil adhesion to biological surfaces (IC50 = 50 nM) as well as adhesion-dependent granule exocytosis and oxidative burst. U-73343 (sc-201422) is useful as a negative control for investigations of U-73122 phospholipase C antagonism and its cellular consequences.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Inhibitor of phospholipase C activation and inhibits the down regulation of muscarinic receptors.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112648-68-7
Molecular Formula C29H40N2O3
Synonym 1-[6-[((17beta)-3-Methoxyestra-1,3,5[10]-trien-17-yl)amino]hexyl]pyrrole-2,5-dione

Specifications

Quantity 1 mg
Description Off-white
For Use With (Application) Inhibitor of phospholipase C activation and inhibits the down regulation of muscarinic receptors
Formula Weight 464.65g/mol
Solubility Soluble in DMSO, ethanol, or DMF at approx. 0.5mg/ml; May require heating or overnight mixing; Insoluble in water
Product Type Signal Transduction Reagent
Form Crystalline solid
Assay ≥99%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.