Learn More
DuPont™ Tyvek™ APX™ 400 TA198S
Product Code. 31903161 Shop All DuPont Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31903161 Medium Pack of 25
31903165 Small Pack of 25
31903159 Large Pack of 25
31903163 X-Large Pack of 25
31903157 2X-Large Pack of 25
31903160 3X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31903161 Supplier DuPont™ Supplier No. D15582417
  • ONLINE EXCLUSIVE PRICE
    £239.67
    £174.68 / Pack of 25
    Save £64.99
    27% Off
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Robust yet lightweight protection providing a barrier against fine particles and low-level sprays in an extremely breathable white hooded coverall made with the innovative new Tyvek™ APX™ fabric.
Features stitched external seams, a respirator fit three-piece hood, thumb loops, an ergonomic design, elasticated wrists, ankles, face, and waist (glued-in) and a Tyvek™ zipper with a pin lock slider zipper pull and a storm flap. Suitable for a variety of applications including pharmaceutical handling, chemical processing, general maintenance, and spray painting.
  • Certified according to Regulation (EU) 2016/425.
  • Chemical protective clothing, Category III, Type 5-B (EN ISO 13982-1) and 6-B (EN 13034).
  • Antistatic treatment (EN 1149-5) on both sides.
  • Barrier to infective agents (EN 14126).
  • Protection against radioactive contamination (EN 1073-2).
  • Tyvek™ thumb loops for improved sleeve fit.
  • Tyvek™ zipper and zipper flap for enhanced protection.
  • Extremely breathable all around.
  • 360° protection and durability of Tyvek™ fabric.
  • Boxes of 25 each without value packs contributing to less plastic waste; individual bags based on post-consumer recycled content.
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Gender Unisex
Material HDPE
Color White
Disposable Single-use
Closure Type Zipper with Storm Flap
Sterility Non-sterile
Resistance Chemicals, Liquid, Particle, Anti-static, Dust
Latex Free Yes
No. of Pockets None