Learn More
DuPont™ Tyvek™ 600 Hooded Coverall, Plus Size
Product Code. 17484943 Shop All DuPont Products
Change view
Click to view available options
Size:
4X-Large
5X-Large
6X-Large
7X-Large
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17484943 4X-Large Pack of 100
17494943 5X-Large Pack of 100
17404953 6X-Large Pack of 100
17414953 7X-Large Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17484943 Supplier DuPont™ Supplier No. D14981422
  • ONLINE EXCLUSIVE PRICE
    £1072.23
    £809.05 / Pack of 100
    Save £263.18
    24% Off
View all promotions
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
Hooded coveralls that are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.
  • Thumb loops
  • Stitched and over-taped seams for protection and strength
  • Tyvek™ zipper and zipper flap for enhanced protection
  • Face, wrist and ankle tunnelled elastication, for good fit and contamination/ linting reduction
TRUSTED_SUSTAINABILITY
Specifications
Size 4X-Large
Color White
Closure Type Zipper
Seam Type Stitched, Taped
Certifications/Compliance (Europe) (EU) 2016/425, EN 14126, EN 1149-5