missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin B42, 99+%, Thermo Scientific™

Product Code. 15444259
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15444259 25 mg 25mg
15454259 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444259 Supplier Thermo Scientific Alfa Aesar Supplier No. J61715.MB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A protein kinase inhibitor

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Tyrphostin B42 is used in acute lymphoblastic leukemia studies the compound has been reported to inhibit JAK2 from B cell precursors.

Solubility
Soluble in DMSO or ethanol

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134036-52-5
Molecular Formula C17H14N2O3
Molecular Weight (g/mol) 294.31
MDL Number MFCD00236452
InChI Key TUCIOBMMDDOEMM-ZSOIEALJSA-N
Synonym 2-cyano-3-3,4-dihydroxyphenyl-n-phenylmethyl-2-propenamide, 2-cyano-3-3,4-dihydroxyphenyl-n-benzyl-2-propenamide, z-n-benzyl-2-cyano-3-3,4-dihydroxyphenyl prop-2-enamide, 2z-n-benzyl-2-cyano-3-3,4-dihydroxyphenyl prop-2-enamide
PubChem CID 5353441
IUPAC Name (Z)-N-benzyl-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enamide
SMILES C1=CC=C(C=C1)CNC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N

Specifications

Melting Point 216°C to 220°C
Color Green-Yellow
Quantity 25 mg
Solubility Information Soluble in DMSO or ethanol
Formula Weight 294.31
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Tyrphostin B42
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.