missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-PEGASUS-90 Auto Closing
Safe and approved storage of flammable hazardous substances in working areas
£2167.17 - £2862.00
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 616 mm |
Depth (Metric) Interior | 524 mm |
Dimensions (D x W x H) | 616 x 596 x 1298 mm |
Dimensions (D x W x H) Interior | 524 x 450 x 1072 mm |
Alert:
For indoor use only
Product Code | Brand | Color | Configuration | Door Style | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Configuration | Door Style | Price | Quantity & Availability | |||
15964046
|
asecos™
3024200133740 |
Light Gray | 2 x drawer (sheet steel powder-coated) | Wing, Left-hinged, Auto-close |
£2275.82
Each |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15924076
|
asecos™
3024200233740 |
Yellow | 2 x drawer (sheet steel powder-coated) | Wing, Left-hinged, Auto-close |
£2275.82
Each |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15302559
|
asecos™
3024200230277 |
Yellow | 2 x shelf, 1 x perforated insert (235 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Wing, Left-hinged, Auto-close |
£2512.20
Each |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15516865
|
asecos™
3024200133735 |
Light Gray | 2 x shelf, 1 x perforated insert (285 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Wing, Left-hinged, Auto-close |
£2167.17
Each |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15974066
|
asecos™
3024200233735 |
Warning Yellow | 2 x shelf, 1 x perforated insert (285 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Wing, Left-hinged, Auto-close |
£2167.17
Each |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15934106
|
asecos™
3024300134413 |
Light Gray | 2 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Wing, Right-hinged, Auto-close |
£2167.17
Each |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15994136
|
asecos™
3024300234413 |
Warning Yellow | 2 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Wing, Right-hinged, Auto-close |
£2167.17
Each |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15974046
|
asecos™
3024200133741 |
Light Gray | 3 x drawer (sheet steel powder-coated) | Wing, Left-hinged, Auto-close |
£2415.74
Each |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15934076
|
asecos™
3024200233741 |
Warning Yellow | 3 x drawer (sheet steel powder-coated) | Wing, Left-hinged, Auto-close |
£2415.74
Each |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15312559
|
asecos™
3024200230722 |
Yellow | 3 x drawer, 1 x bottom collecting sump (sheet steel powder-coated) | Wing, Left-hinged, Auto-close |
£2862.00
Each |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15924046
|
asecos™
3024200133736 |
Light Gray | 3 x shelf, 1 x perforated insert (185 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Wing, Left-hinged, Auto-close |
£2204.80
Each |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15984066
|
asecos™
3024200233736 |
Warning Yellow | 3 x shelf, 1 x perforated insert (185 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Wing, Left-hinged, Auto-close |
£2204.80
Each |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15944106
|
asecos™
3024300134414 |
Light Gray | 3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Wing, Right-hinged, Auto-close |
£2204.80
Each |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15904146
|
asecos™
3024300234414 |
Yellow | 3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Wing, Right-hinged, Auto-close |
£2204.80
Each |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- Always closed: serial automatic door closing (TSA) releases arrested door after approx. 60 seconds; acoustic and visual signal before closing
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
524 mm | |
524 x 450 x 1072 mm | |
Safety storage cabinet | |
1298 mm | |
1 | |
596 mm |
616 mm | |
616 x 596 x 1298 mm | |
Steel | |
Safe and approved storage of flammable hazardous substances in working areas | |
1072 mm | |
Powder-coated plain | |
450 mm |